ENA346975015 Enamine

1-(cyclopropylmethyl)-1H-pyrazol-4-amine

  • MDL number MFCD12138807
  • Cas Number : 1156169-29-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95597

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-95597
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart