ENA346975020 Enamine

4-(1,3-dioxolan-2-yl)-4-methylheptan-1-amine

  • MDL number MFCD20731178
  • Cas Number : 1354963-75-9
  • Molecular Weight : 201.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-95603

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-95603
Availability details for
Quantity
  • Availability
  • Add to Cart