ENA346975022 Enamine

2-phenyl-1-(pyridin-4-yl)ethan-1-one

  • MDL number MFCD12153293
  • Cas Number : 1017-24-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95613

Safety Information

msds https://www.enaminestore.com/catalog/EN300-95613
Availability details for
Quantity
  • Availability
  • Add to Cart