ENA346975027 Enamine

ethyl({[1-(pyridin-2-yl)piperidin-4-yl]methyl})amine

  • MDL number MFCD16806408
  • Cas Number : 1354961-36-6
  • Molecular Weight : 219.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95618

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-95618
Availability details for
Quantity
  • Availability
  • Add to Cart