ENA346975049 Enamine

2-methyl-5-phenylmorpholine

  • MDL number MFCD18285361
  • Cas Number : 1343334-74-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95674

Safety Information

msds https://www.enaminestore.com/catalog/EN300-95674
Availability details for
Quantity
  • Availability
  • Add to Cart