ENA346975050 Enamine

2-[(4-sulfamoylphenyl)sulfanyl]benzoic acid

  • MDL number MFCD13699562
  • Cas Number : 1154996-56-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 226...228
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85790

Safety Information

msds https://www.enaminestore.com/catalog/EN300-85790
Availability details for
Quantity
  • Availability
  • Add to Cart