ENA346975063 Enamine

3,3,3-trifluoropropanethioamide

  • MDL number MFCD09804631
  • Cas Number : 365428-34-8
  • Molecular Weight : 143.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-95696

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-95696
Availability details for
Quantity
  • Availability
  • Add to Cart