ENA346975065 Enamine

2-(3-nitrophenoxy)-2-phenylacetic acid

  • MDL number MFCD09721280
  • Cas Number : 938303-74-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95701

Safety Information

msds https://www.enaminestore.com/catalog/EN300-95701
Availability details for
Quantity
  • Availability
  • Add to Cart