ENA346975070 Enamine

2-ethyl-2-phenylbutanoic acid

  • MDL number MFCD00075036
  • Cas Number : 5465-28-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95752

Safety Information

msds https://www.enaminestore.com/catalog/EN300-95752
Availability details for
Quantity
  • Availability
  • Add to Cart