ENA346975076 Enamine

ethyl 3-hydroxy-3-methylbutanoate

  • MDL number MFCD16749012
  • Cas Number : 18267-36-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95759

Safety Information

msds https://www.enaminestore.com/catalog/EN300-95759
Availability details for
Quantity
  • Availability
  • Add to Cart