ENA346975077 Enamine

1-benzyl-4-methyl-1H-pyrazol-3-amine

  • MDL number MFCD11215421
  • Cas Number : 1174838-25-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-95760

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-95760
Availability details for
Quantity
  • Availability
  • Add to Cart