ENA346975081 Enamine

5-chloro-3-(propan-2-yl)-1,2,4-thiadiazole

  • MDL number MFCD09743916
  • Cas Number : 51302-12-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-87249

Safety Information

msds https://www.enaminestore.com/catalog/EN300-87249
Availability details for
Quantity
  • Availability
  • Add to Cart