ENA346975090 Enamine

2-(5-phenyl-2H-1,2,3,4-tetrazol-2-yl)propanoic acid

  • MDL number MFCD01871021
  • Cas Number : 22307-40-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 143...145
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95789

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-95789
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart