ENA346975091 Enamine

2-ethoxy-3-fluorobenzoic acid

  • MDL number MFCD16836018
  • Cas Number : 1248787-46-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95794

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-95794
Availability details for
Quantity
  • Availability
  • Add to Cart