ENA346975093 Enamine

4-(aminomethyl)-N,N-dimethylquinolin-2-amine

  • MDL number MFCD16703574
  • Cas Number : 1247541-98-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95798

Safety Information

msds https://www.enaminestore.com/catalog/EN300-95798
Availability details for
Quantity
  • Availability
  • Add to Cart