ENA346975107 Enamine

methyl 4-methoxythiophene-2-carboxylate

  • MDL number MFCD17012823
  • Cas Number : 5118-05-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95869

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-95869
Availability details for
Quantity
  • Availability
  • Add to Cart