ENA346975110 Enamine

4-amino-3-fluorobenzamide

  • MDL number MFCD16658741
  • Cas Number : 609783-62-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 152...154
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95896

Safety Information

msds https://www.enaminestore.com/catalog/EN300-95896
Availability details for
Quantity
  • Availability
  • Add to Cart