ENA346975111 Enamine

2-chloro-6-(chloromethyl)pyridine

  • MDL number MFCD07774109
  • Cas Number : 78846-88-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 42...44
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95897

Safety Information

msds https://www.enaminestore.com/catalog/EN300-95897
Availability details for
Quantity
  • Availability
  • Add to Cart