ENA346975116 Enamine

4-{6-methyl-[1,2,4]triazolo[4,3-b]pyridazin-3-yl}piperidine

  • MDL number MFCD16169043
  • Cas Number : 1249085-98-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95905

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-95905
Availability details for
Quantity
  • Availability
  • Add to Cart