ENA346975118 Enamine

N-[(4-nitrophenyl)methyl]aniline

  • MDL number MFCD00439160
  • Cas Number : 10359-18-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95908

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-95908
Availability details for
Quantity
  • Availability
  • Add to Cart