ENA346975120 Enamine

methyl 3-{[(2,3-difluorophenyl)methyl]amino}propanoate

  • MDL number MFCD12762129
  • Cas Number : 1183882-14-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-95913
Availability details for
Quantity
  • Availability
  • Add to Cart