ENA346975124 Enamine

1-(4-methylphenyl)-2-(pyridin-2-yl)ethan-1-one

  • MDL number MFCD11188614
  • Cas Number : 72076-59-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 73...75
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95943

Safety Information

msds https://www.enaminestore.com/catalog/EN300-95943
Availability details for
Quantity
  • Availability
  • Add to Cart