ENA346975138 Enamine

1-(4-fluorophenyl)-2-methoxyethan-1-amine hydrochloride

  • MDL number MFCD20731200
  • Cas Number : 1354963-82-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-95994

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-95994
Availability details for
Quantity
  • Availability
  • Add to Cart