ENA346975139 Enamine

4-(pyrazin-2-yloxy)benzaldehyde

  • MDL number MFCD02667635
  • Cas Number : 866156-93-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-95995

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-95995
Availability details for
Quantity
  • Availability
  • Add to Cart