ENA346975143 Enamine

N-(4-methoxy-2-nitrophenyl)prop-2-enamide

  • MDL number MFCD12091404
  • Cas Number : 444166-94-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96002

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96002
Availability details for
Quantity
  • Availability
  • Add to Cart