ENA346975144 Enamine

bis(prop-2-yn-1-yl)amine hydrochloride

  • MDL number MFCD11109425
  • Cas Number : 93282-90-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 187...189
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96003

Safety Information

msds https://www.enaminestore.com/catalog/EN300-96003
Availability details for
Quantity
  • Availability
  • Add to Cart