ENA346975153 Enamine

2,2,2-trifluoro-1-[3-(trifluoromethyl)phenyl]ethan-1-ol

  • MDL number MFCD00455619
  • Cas Number : 721-36-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 31...33
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-90720

Safety Information

msds https://www.enaminestore.com/catalog/EN300-90720
Availability details for
Quantity
  • Availability
  • Add to Cart