ENA346975161 Enamine

methyl 2-oxo-2,3-dihydro-1H-1,3-benzodiazole-5-carboxylate

  • MDL number MFCD08457979
  • Cas Number : 106429-57-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96163

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96163
Availability details for
Quantity
  • Availability
  • Add to Cart