ENA346975178 Enamine

7-bromo-1,2-benzoxazole

  • MDL number MFCD11520894
  • Cas Number : 1260751-81-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 79...81
Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96187

Safety Information

msds https://www.enaminestore.com/catalog/EN300-96187
Availability details for
Quantity
  • Availability
  • Add to Cart