ENA346975179 Enamine

2-[cyclopentyl(methyl)amino]pyridine-4-carboxylic acid

  • MDL number MFCD11618576
  • Cas Number : 1094661-79-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96188

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96188
Availability details for
Quantity
  • Availability
  • Add to Cart