ENA346975187 Enamine

1-methyl-3-(piperazin-1-yl)-1,2-dihydropyrazin-2-one

  • MDL number MFCD17249972
  • Cas Number : 1343640-83-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96206

Safety Information

msds https://www.enaminestore.com/catalog/EN300-96206
Availability details for
Quantity
  • Availability
  • Add to Cart