ENA346975188 Enamine

2-cyclopropyl-2-hydroxyacetonitrile

  • MDL number MFCD14622836
  • Cas Number : 5648-87-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96207

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96207
Availability details for
Quantity
  • Availability
  • Add to Cart