ENA346975189 Enamine

2-(3-bromophenyl)-1,1,1-trifluoropropan-2-ol

  • MDL number MFCD16068003
  • Cas Number : 122243-30-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96208

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96208
Availability details for
Quantity
  • Availability
  • Add to Cart