ENA346975205 Enamine

1-methyl-3-(pyridin-4-yl)-1H-pyrazol-4-amine

  • MDL number MFCD20621146
  • Cas Number : 1313013-53-4
  • Molecular Weight : 174.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-91703

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-91703
Availability details for
Quantity
  • Availability
  • Add to Cart