ENA346975218 Enamine

1-(phthalazin-1-yl)piperidin-4-ol

  • MDL number MFCD13196937
  • Cas Number : 1308246-46-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135...137
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96283

Safety Information

msds https://www.enaminestore.com/catalog/EN300-96283
Availability details for
Quantity
  • Availability
  • Add to Cart