ENA346975219 Enamine

2-(piperidin-4-yl)-5-(trifluoromethyl)aniline

  • MDL number MFCD17486505
  • Cas Number : 1260823-98-0
  • Molecular Weight : 244.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96284

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96284
Availability details for
Quantity
  • Availability
  • Add to Cart