ENA346975220 Enamine

tert-butyl N-(1,3-diaminopropan-2-yl)carbamate

  • MDL number MFCD20696567
  • Cas Number : 213475-74-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96285

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96285
Availability details for
Quantity
  • Availability
  • Add to Cart