ENA346975221 Enamine

6-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine

  • MDL number MFCD13182009
  • Cas Number : 1220219-97-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96286

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96286
Availability details for
Quantity
  • Availability
  • Add to Cart