ENA346975233 Enamine

1,2,3-trichloroprop-1-ene

  • Cas Number : 96-19-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Flash Point 82
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96344

Safety Information

signalword Danger
pcodes P210,P261,P264,P270,P271,P280,P301+P310,P302+P352,P304+P340,P311,P312,P330,P361,P363,P370+P378,P403+P233,P403+P235,P405,P501
hcodes T
MSDS https://www.enaminestore.com/catalog/EN300-96344
pictograms GHS06
hstatements H227,H301,H311,H331
Availability details for
Quantity
  • Availability
  • Add to Cart