ENA346975241 Enamine

2-(cyclopropylformamido)-3-hydroxybutanoic acid

  • MDL number MFCD09836924
  • Cas Number : 1160934-95-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96362

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96362
Availability details for
Quantity
  • Availability
  • Add to Cart