ENA346975258 Enamine

1-(2-chlorophenyl)-1H-pyrazol-3-amine

  • MDL number MFCD16845375
  • Cas Number : 76091-02-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92002

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92002
Availability details for
Quantity
  • Availability
  • Add to Cart