ENA346975261 Enamine

9-methyl-3,9-diazabicyclo[4.2.1]nonane

  • MDL number MFCD13192515
  • Cas Number : 102547-84-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96392

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96392
Availability details for
Quantity
  • Availability
  • Add to Cart