ENA346975268 Enamine

1-sulfanylpropan-2-one

  • MDL number MFCD00129680
  • Cas Number : 24653-75-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 109-111
Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-92052

Safety Information

signalword Danger
pcodes P210,P240,P241,P264,P270,P280,P301+P312,P305+P351+P338,P310,P330,P370+P378,P501
hcodes Xi,Xn
MSDS https://www.enaminestore.com/catalog/EN300-92052
pictograms GHS02,GHS05,GHS07
hstatements H228,H302,H318
Availability details for
Quantity
  • Availability
  • Add to Cart