ENA346975275 Enamine

N-methyl-2-(piperidin-4-yloxy)acetamide

  • MDL number MFCD08061037
  • Cas Number : 912761-44-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96412

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96412
Availability details for
Quantity
  • Availability
  • Add to Cart