ENA346975276 Enamine

(4-ethyloxan-4-yl)methanamine

  • MDL number MFCD12028172
  • Cas Number : 1142202-08-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96414

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96414
Availability details for
Quantity
  • Availability
  • Add to Cart