ENA346975279 Enamine

methyl 1-{thieno[3,2-c]pyridin-4-yl}piperidine-4-carboxylate

  • MDL number MFCD15486653
  • Cas Number : 1354954-10-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96417

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96417
Availability details for
Quantity
  • Availability
  • Add to Cart