ENA346975280 Enamine

2-(cyclopentylamino)pyridine-4-carboxylic acid

  • MDL number MFCD11132015
  • Cas Number : 1019127-19-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-96421

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96421
Availability details for
Quantity
  • Availability
  • Add to Cart