ENA346975282 Enamine

2-tert-butyl-4-chloropyrimidine

  • MDL number MFCD10697056
  • Cas Number : 18436-67-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93137

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93137
Availability details for
Quantity
  • Availability
  • Add to Cart