ENA346975284 Enamine

methyl 4-(chlorosulfonyl)-1-methyl-1H-pyrrole-2-carboxylate

  • MDL number MFCD14281198
  • Cas Number : 69812-32-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 78...80
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96474

Safety Information

msds https://www.enaminestore.com/catalog/EN300-96474
Availability details for
Quantity
  • Availability
  • Add to Cart