ENA346975285 Enamine

4-amino-3-chlorobenzene-1-sulfonamide

  • MDL number MFCD01565841
  • Cas Number : 53297-68-0
  • Molecular Weight : 206.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-96476

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96476
Availability details for
Quantity
  • Availability
  • Add to Cart